(S)-(+)-2-Phenylglycine methyl ester hydrochloride 97%

BD689134-25G BD689134-100G
BLDPBD689134-25GEA 16.8 EUR
BLDPBD689134-25G BLDPBD689134-100G
(S)-(+)-2-Phenylglycine methyl ester hydrochloride 97%
(S)-(+)-2-Phenylglycine methyl ester hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formula: C₉H₁₂ClNO₂
MW: 201.65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00077158
CAS Number: 15028-39-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR