2-Ethoxy-benzamidine hydrochloride 96%
|
|
BD6895-5G
BD6895-1G
BD6895-250MG
BLDPBD6895-5GEA
26.3
EUR
BLDPBD6895-5G
BLDPBD6895-1G
BLDPBD6895-250MG
2-Ethoxy-benzamidine hydrochloride 96%
2-Ethoxy-benzamidine hydrochloride
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₉H₁₂N₂O·ClH Storage Temperature: Ambient |
MDL Number:
MFCD05663676 CAS Number: 18637-00-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...