Ethyl 2,4-dimethylpyrrole-3-carboxylate 97%

BD6968-100G BD6968-250MG
BLDPBD6968-100GEA 565 EUR
BLDPBD6968-100G BLDPBD6968-250MG
Ethyl 2,4-dimethylpyrrole-3-carboxylate 97%
Ethyl 2,4-dimethylpyrrole-3-carboxylate

Storage condition: Inert atmosphere, Room Temperature

Formula: C₉H₁₃NO₂
MW: 167.2050 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01365811
CAS Number: 2199-51-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR