2-Amino-4-methyl-5-pyridinecarboxylic acid 95%

BD70220-5G BD70220-100MG
BLDPBD70220-5GEA 306 EUR
BLDPBD70220-5G BLDPBD70220-100MG
2-Amino-4-methyl-5-pyridinecarboxylic acid 95%
2-Amino-4-methyl-5-pyridinecarboxylic acid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₇H₈N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD09835242
CAS Number: 179555-11-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR