Boc-Thz-OH 98%

BD7050-25G BD7050-100G
BLDPBD7050-25GEA 50.4 EUR
BLDPBD7050-25G BLDPBD7050-100G
Boc-Thz-OH 98%
Boc-Thz-OH

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₁₅NO₄S
MW: 233.282 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00077002
CAS Number: 51077-16-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR