4,5,6,7-Tetrahydrobenzo[b]thiophene-2-carboxamide 97%

BD707235-1G BD707235-250MG
BLDPBD707235-1GEA 483 EUR
BLDPBD707235-1G BLDPBD707235-250MG
4,5,6,7-Tetrahydrobenzo[b]thiophene-2-carboxamide 97%
4,5,6,7-Tetrahydrobenzo[b]thiophene-2-carboxamide

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₉H₁₁NOS
Storage Temperature: Ambient
MDL Number: MFCD00460789
CAS Number: 57021-58-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR