2-Aminobenzenesulphonic acid 98%

BD70733-500G BD70733-100G
BLDPBD70733-500GEA 38.9 EUR
BLDPBD70733-500G BLDPBD70733-100G
2-Aminobenzenesulphonic acid 98%
2-Aminobenzenesulphonic acid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₆H₇NO₃S
Storage Temperature: Ambient
MDL Number: MFCD00007705
CAS Number: 88-21-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR