2-Amino-4-ethoxybenzoic acid 97%

BD708810-1G BD708810-250MG
BLDPBD708810-1GEA 265 EUR
BLDPBD708810-1G BLDPBD708810-250MG
2-Amino-4-ethoxybenzoic acid 97%
2-Amino-4-ethoxybenzoic acid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₉H₁₁NO₃
Storage Temperature: Ambient
MDL Number: MFCD01464112
CAS Number: 61484-98-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR