4-Amino-N-phenylbenzenesulphonamide 98%

BD70908-1G BD70908-5G
BLDPBD70908-1GEA 124 EUR
BLDPBD70908-1G BLDPBD70908-5G
4-Amino-N-phenylbenzenesulphonamide 98%
4-Amino-N-phenylbenzenesulphonamide

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₁₂H₁₂N₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD00233654
CAS Number: 127-77-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR