4-Chloro-6-fluoroquinoline 97%

BD7187-5G BD7187-10G
BLDPBD7187-5GEA 25.2 EUR
BLDPBD7187-5G BLDPBD7187-10G
4-Chloro-6-fluoroquinoline 97%
4-Chloro-6-fluoroquinoline

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₉H₅ClFN
Storage Temperature: Ambient
MDL Number: MFCD00278783
CAS Number: 391-77-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR