2-Bromo-4-tert-butylphenol 98%
|
|
BD71877-10G
BD71877-5G
BLDPBD71877-10GEA
11.6
EUR
BLDPBD71877-10G
BLDPBD71877-5G
2-Bromo-4-tert-butylphenol 98%
2-Bromo-4-tert-butylphenol
Storage condition: Sealed in dry, Room Temperature
Formula:
C₁₀H₁₃BrO Storage Temperature: Ambient |
MDL Number:
MFCD02682891 CAS Number: 2198-66-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...