2-(Carboxymethyl)-1-methyl-1H-pyrrole-3-carboxylic acid 98%

BD72460-1G BD72460-250MG
BLDPBD72460-1GEA 68.3 EUR
BLDPBD72460-1G BLDPBD72460-250MG
2-(Carboxymethyl)-1-methyl-1H-pyrrole-3-carboxylic acid 98%
2-(Carboxymethyl)-1-methyl-1H-pyrrole-3-carboxylic acid
Formula: C₈H₉NO₄
MDL Number: MFCD00044904
CAS Number: 83863-74-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR