2-Chloro-6-methoxypyrazine 98%

BD73401-500G BD73401-5G
BLDPBD73401-500GEA 1820 EUR
BLDPBD73401-500G BLDPBD73401-5G
2-Chloro-6-methoxypyrazine 98%
2-Chloro-6-methoxypyrazine

Storage condition: Inert atmosphere, 2-8°C

Formula: C₅H₅ClN₂O
MW: 144.5600 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06410937
CAS Number: 33332-30-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR