2-Chloro-6-methylquinoline 98%
|
|
BD73444-1G
BD73444-250MG
BLDPBD73444-1GEA
50.4
EUR
BLDPBD73444-1G
BLDPBD73444-250MG
2-Chloro-6-methylquinoline 98%
2-Chloro-6-methylquinoline
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₁₀H₈ClN Storage Temperature: Ambient |
MDL Number:
MFCD00462389 CAS Number: 4295-11-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...