2'-Chloro-4'-(trifluoromethyl)acetanilide 97%
|
|
BD7377-25G
BD7377-5G
BLDPBD7377-25GEA
108
EUR
BLDPBD7377-25G
BLDPBD7377-5G
2'-Chloro-4'-(trifluoromethyl)acetanilide 97%
2'-Chloro-4'-(trifluoromethyl)acetanilide
Storage condition: Sealed in dry, Room Temperature
Formula:
C₉H₇ClF₃NO Storage Temperature: Ambient |
MDL Number:
MFCD01313231 CAS Number: 247170-19-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...