3,4-Difluoro-N-methylaniline 97%

BD749043-5G BD749043-25G
BLDPBD749043-5GEA 425 EUR
BLDPBD749043-5G BLDPBD749043-25G
3,4-Difluoro-N-methylaniline 97%
3,4-Difluoro-N-methylaniline

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₇H₇F₂N
MW: 143.13 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11152726
CAS Number: 138563-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR