Ethyl 5-bromo-6-chloro-1H-indole-2-carboxylate 95%

BD750987-5G BD750987-1G
BLDPBD750987-5GEA 1010 EUR
BLDPBD750987-5G BLDPBD750987-1G
Ethyl 5-bromo-6-chloro-1H-indole-2-carboxylate 95%
Ethyl 5-bromo-6-chloro-1H-indole-2-carboxylate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₁H₉BrClNO₂
Storage Temperature: Ambient
MDL Number: MFCD21363140
CAS Number: 1923237-13-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR