Thiazolo[4,5-d]pyrimidin-7(6H)-one 97%

BD751412-50MG BD751412-100MG
BLDPBD751412-50MGEA 128 EUR
BLDPBD751412-50MG BLDPBD751412-100MG
Thiazolo[4,5-d]pyrimidin-7(6H)-one 97%
Thiazolo[4,5-d]pyrimidin-7(6H)-one

Storage condition: Sealed in dry, Room Temperature

Formula: C₅H₃N₃OS
Storage Temperature: Ambient
MDL Number: MFCD30188564
CAS Number: 1353101-44-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR