N-(2-Aminoethyl)-1,2,3,4-tetrahydronaphthalene-1-carboxamide hydrochloride 97%

BD751601-100MG BD751601-250MG
BLDPBD751601-100MGEA 108 EUR
BLDPBD751601-100MG BLDPBD751601-250MG
N-(2-Aminoethyl)-1,2,3,4-tetrahydronaphthalene-1-carboxamide hydrochloride 97%
N-(2-Aminoethyl)-1,2,3,4-tetrahydronaphthalene-1-carboxamide hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₃H₁₉ClN₂O
Storage Temperature: Ambient
MDL Number: MFCD27966972
CAS Number: 1828966-95-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR