2-Fluoroisonicotinonitrile 98%

BD75161-5G BD75161-500G
BLDPBD75161-5GEA 24.2 EUR
BLDPBD75161-5G BLDPBD75161-500G
2-Fluoroisonicotinonitrile 98%
2-Fluoroisonicotinonitrile

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₆H₃FN₂
Storage Temperature: Ambient
MDL Number: MFCD09027299
CAS Number: 3939-14-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR