6-Chloro-4-ethoxypyridin-3-amine 95%
|
|
BD751687-1G
BD751687-5G
BLDPBD751687-1GEA
525
EUR
BLDPBD751687-1G
BLDPBD751687-5G
6-Chloro-4-ethoxypyridin-3-amine 95%
6-Chloro-4-ethoxypyridin-3-amine
Storage condition: Keep in dark place, Inert atmosphere, 2-8°C
Formula:
C₇H₉ClN₂O Storage Temperature: Refrigerator |
MDL Number:
MFCD28666482 CAS Number: 872282-68-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...