3-Bromo-4-methylpyridin-2-ol 98%

BD75542-25G BD75542-100MG
BLDPBD75542-25GEA 433 EUR
BLDPBD75542-25G BLDPBD75542-100MG
3-Bromo-4-methylpyridin-2-ol 98%
3-Bromo-4-methylpyridin-2-ol

Storage condition: Sealed in dry, Room Temperature

Formula: C₆H₆BrNO
Storage Temperature: Ambient
MDL Number: MFCD09839708
CAS Number: 18368-59-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR