Bis(2,2'-bipyridine-κN1,κN1')(dipyrido[3,2-a:2',3'-c]phenazine-κN4,κN5)ruthenium(2+) 98%
|
|
BD762213-50MG
BD762213-25MG
BLDPBD762213-50MGEA
322
EUR
BLDPBD762213-50MG
BLDPBD762213-25MG
Bis(2,2'-bipyridine-κN1,κN1')(dipyrido[3,2-a:2',3'-c]phenazine-κN4,κN5)ruthenium(2+) 98%
Bis(2,2'-bipyridine-κN1,κN1')(dipyrido[3,2-a:2',3'-c]phenazine-κN4,κN5)ruthenium(2+)
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formula:
C₃₈H₂₆N₈Ru MW: 695.73 g/mol Storage Temperature: Ambient |
CAS Number:
87564-74-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...