(4-(Morpholinomethyl)phenyl)methanamine 95%
|
|
BD768594-5G
BD768594-1G
BLDPBD768594-5GEA
210
EUR
BLDPBD768594-5G
BLDPBD768594-1G
(4-(Morpholinomethyl)phenyl)methanamine 95%
(4-(Morpholinomethyl)phenyl)methanamine
Storage condition: Keep in dark place, Sealed in dry, 2-8°C
Formula:
C₁₂H₁₈N₂O Storage Temperature: Refrigerator |
MDL Number:
MFCD05664410 CAS Number: 91271-84-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...