2-Methyl-3-nitrobenzonitrile 98%
|
|
BD76868-25G
BD76868-5G
BLDPBD76868-25GEA
222
EUR
BLDPBD76868-25G
BLDPBD76868-5G
2-Methyl-3-nitrobenzonitrile 98%
2-Methyl-3-nitrobenzonitrile
Storage condition: Sealed in dry, Room Temperature
Formula:
C₈H₆N₂O₂ MW: 162.15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09025714 CAS Number: 71516-35-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...