5-Nitrothiophene-3-carbaldehyde 98%

BD77789-25G BD77789-5G
BLDPBD77789-25GEA 853 EUR
BLDPBD77789-25G BLDPBD77789-5G
5-Nitrothiophene-3-carbaldehyde 98%
5-Nitrothiophene-3-carbaldehyde

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₅H₃NO₃S
Storage Temperature: Refrigerator
MDL Number: MFCD00052581
CAS Number: 75428-45-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR