2-(1-Pyrazolyl)pyridine 98%
|
|
BD78444-5G
BD78444-1G
BLDPBD78444-5GEA
182
EUR
BLDPBD78444-5G
BLDPBD78444-1G
2-(1-Pyrazolyl)pyridine 98%
2-(1-Pyrazolyl)pyridine
Storage condition: Sealed in dry, Room Temperature
Formula:
C₈H₇N₃ Storage Temperature: Ambient |
MDL Number:
MFCD09033534 CAS Number: 25700-11-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...