2-(tert-Butyl)-1H-indole 97%
|
|
BD78665-5G
BD78665-25G
BLDPBD78665-5GEA
310
EUR
BLDPBD78665-5G
BLDPBD78665-25G
2-(tert-Butyl)-1H-indole 97%
2-(tert-Butyl)-1H-indole
Storage condition: Keep in dark place, Sealed in dry, 2-8°C
Formula:
C₁₂H₁₅N Storage Temperature: Refrigerator |
MDL Number:
MFCD04004060 CAS Number: 1805-65-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...