Boc-Tic-OH 98%

BD7870-500G BD7870-25G
BLDPBD7870-500GEA 559 EUR
BLDPBD7870-500G BLDPBD7870-25G
Boc-Tic-OH 98%
Boc-Tic-OH

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₅H₁₉NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD00143845
CAS Number: 78879-20-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR