(2R,3R,4S,5R)-2-(2,6-Dichloro-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol 97%

BD7936-250MG BD7936-1G
BLDPBD7936-250MGEA 52.5 EUR
BLDPBD7936-250MG BLDPBD7936-1G
(2R,3R,4S,5R)-2-(2,6-Dichloro-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol 97%
(2R,3R,4S,5R)-2-(2,6-Dichloro-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₀H₁₀Cl₂N₄O₄
MW: 321.12 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03411342
CAS Number: 13276-52-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR