3-(Carboxymethyl)benzothiazolium bromide 97%

BD80519-100G BD80519-1G
BLDPBD80519-100GEA 330 EUR
BLDPBD80519-100G BLDPBD80519-1G
3-(Carboxymethyl)benzothiazolium bromide 97%
3-(Carboxymethyl)benzothiazolium bromide

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₈BrNO₂S
MW: 274.1343 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00134360
CAS Number: 74385-09-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR