2'-Deoxy-2'-fluorouridine 98%

BD8058-250MG BD8058-5G
BLDPBD8058-250MGEA 9.5 EUR
BLDPBD8058-250MG BLDPBD8058-5G
2'-Deoxy-2'-fluorouridine 98%
2'-Deoxy-2'-fluorouridine

Storage condition: Inert atmosphere, 2-8°C

Formula: C₉H₁₁FN₂O₅
MW: 246.19 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01317293
CAS Number: 784-71-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR