3-Dimethylamino-1-pyridin-2-ylpropenone 97%

BD80636-5G BD80636-25G
BLDPBD80636-5GEA 27.3 EUR
BLDPBD80636-5G BLDPBD80636-25G
3-Dimethylamino-1-pyridin-2-ylpropenone 97%
3-Dimethylamino-1-pyridin-2-ylpropenone

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₀H₁₂N₂O
MW: 176.22 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00115039
CAS Number: 66521-54-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR