3-(Dimethylamino)benzaldehyde 97%

BD80652-5G BD80652-1G
BLDPBD80652-5GEA 1110 EUR
BLDPBD80652-5G BLDPBD80652-1G
3-(Dimethylamino)benzaldehyde 97%
3-(Dimethylamino)benzaldehyde

Storage condition: Inert atmosphere, 2-8°C

Formula: C₉H₁₁NO
MW: 149.1897 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD05148649
CAS Number: 619-22-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR