3-Aminoisonicotinic acid 97% (contains <5%AcOH

BD8140-10G BD8140-100G
BLDPBD8140-10GEA 42 EUR
BLDPBD8140-10G BLDPBD8140-100G
3-Aminoisonicotinic acid 97% (contains <5%AcOH
3-Aminoisonicotinic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₆H₆N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00137840
CAS Number: 7579-20-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR