3-Methoxy-4-methylbenzaldehyde 98%

BD8190-1G BD8190-25G
BLDPBD8190-1GEA 24.2 EUR
BLDPBD8190-1G BLDPBD8190-25G
3-Methoxy-4-methylbenzaldehyde 98%
3-Methoxy-4-methylbenzaldehyde

Storage condition: Inert atmosphere, Room Temperature

Formula: C9H10O2
Storage Temperature: Ambient
MDL Number: MFCD04114050
CAS Number: 24973-22-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR