3-Acetyl-6-bromoindole 95%
|
|
BD83551-250MG
BD83551-100MG
BLDPBD83551-250MGEA
186
EUR
BLDPBD83551-250MG
BLDPBD83551-100MG
3-Acetyl-6-bromoindole 95%
3-Acetyl-6-bromoindole
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formula:
C₁₀H₈BrNO Storage Temperature: Ambient |
MDL Number:
MFCD09834653 CAS Number: 316181-82-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...