5-Nitroindole 97%

BD8425-10G BD8425-100G
BLDPBD8425-10GEA 20 EUR
BLDPBD8425-10G BLDPBD8425-100G
5-Nitroindole 97%
5-Nitroindole

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₈H₆N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00005673
CAS Number: 6146-52-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR