3-Aminophenethyl alcohol 97%

BD84537-10G BD84537-100MG
BLDPBD84537-10GEA 535 EUR
BLDPBD84537-10G BLDPBD84537-100MG
3-Aminophenethyl alcohol 97%
3-Aminophenethyl alcohol

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₈H₁₁NO
Storage Temperature: Ambient
MDL Number: MFCD02683213
CAS Number: 52273-77-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR