5-Benzyl-N-(tert-Butoxycarbonyl)-L-glutamate 98%

BD8514-100G BD8514-10G
BLDPBD8514-100GEA 79.8 EUR
BLDPBD8514-100G BLDPBD8514-10G
5-Benzyl-N-(tert-Butoxycarbonyl)-L-glutamate 98%
5-Benzyl-N-(tert-Butoxycarbonyl)-L-glutamate

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₁₇H₂₃NO₆
Storage Temperature: Ambient
MDL Number: MFCD00065569
CAS Number: 13574-13-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR