Boc-L-leucine monohydrate 95%
|
|
BD8523-100G
BD8523-5G
BD8523-10G
BLDPBD8523-100GEA
35.7
EUR
BLDPBD8523-100G
BLDPBD8523-5G
BLDPBD8523-10G
Boc-L-leucine monohydrate 95%
Boc-L-leucine monohydrate
Storage condition: Sealed in dry, Room Temperature
Formula:
C₁₁H₂₁NO₄·H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00065582 CAS Number: 200936-87-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...