3-Bromobenzeneboronic acid-N-methyldiethanolamine ester 98+%

BD85286-5G BD85286-1G
BLDPBD85286-5GEA 159 EUR
BLDPBD85286-5G BLDPBD85286-1G
3-Bromobenzeneboronic acid-N-methyldiethanolamine ester 98+%
3-Bromobenzeneboronic acid-N-methyldiethanolamine ester

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₁H₁₅BBrNO₂
Storage Temperature: Ambient
MDL Number: MFCD02093962
CAS Number: 374538-00-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR