Cyclopropylboronic acid pinacol ester 97%

BD8567-1G BD8567-25G
BLDPBD8567-1GEA 10.5 EUR
BLDPBD8567-1G BLDPBD8567-25G
Cyclopropylboronic acid pinacol ester 97%
Cyclopropylboronic acid pinacol ester

Storage condition: Inert atmosphere, Room Temperature

Formula: C₉H₁₇BO₂
Storage Temperature: Ambient
MDL Number: MFCD05663847
CAS Number: 126689-01-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR