3-Methylbenzofuran 98%

BD88164-25G BD88164-250MG BD88164-10G
BLDPBD88164-25GEA 901 EUR
BLDPBD88164-25G BLDPBD88164-250MG BLDPBD88164-10G
3-Methylbenzofuran 98%
3-Methylbenzofuran

Storage condition: Sealed in dry, 2-8°C

Formula: C₉H₈O
Storage Temperature: Refrigerator
MDL Number: MFCD00205641
CAS Number: 21535-97-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR