3-(tert-Butoxycarbonyl)phenylboronic acid pinacol ester 97%

BD89214-25G BD89214-1G
BLDPBD89214-25GEA 203 EUR
BLDPBD89214-25G BLDPBD89214-1G
3-(tert-Butoxycarbonyl)phenylboronic acid pinacol ester 97%
3-(tert-Butoxycarbonyl)phenylboronic acid pinacol ester

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₇H₂₅BO₄
Storage Temperature: Refrigerator
MDL Number: MFCD03453057
CAS Number: 903895-48-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR