4-(2-Aminopyrimidin-5-yl)benzoic acid 98%

BD89786-250MG BD89786-1G
BLDPBD89786-250MGEA 31.5 EUR
BLDPBD89786-250MG BLDPBD89786-1G
4-(2-Aminopyrimidin-5-yl)benzoic acid 98%
4-(2-Aminopyrimidin-5-yl)benzoic acid

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₁H₉N₃O₂
MW: 215.2100 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD05864814
CAS Number: 222987-21-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR