2-Butyl-3-(4-hydroxy-3,5-diiodobenzoyl)benzofuran 98%
|
|
BD9018-5G
BD9018-250MG
BLDPBD9018-5GEA
636
EUR
BLDPBD9018-5G
BLDPBD9018-250MG
2-Butyl-3-(4-hydroxy-3,5-diiodobenzoyl)benzofuran 98%
2-Butyl-3-(4-hydroxy-3,5-diiodobenzoyl)benzofuran
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formula:
C₁₉H₁₆I₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD02675787 CAS Number: 1951-26-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...