4-Cyano-4'-hexylbiphenyl 98% (by GC)
|
|
BD9041-250MG
BD9041-100MG
BLDPBD9041-250MGEA
31.5
EUR
BLDPBD9041-250MG
BLDPBD9041-100MG
4-Cyano-4'-hexylbiphenyl 98% (by GC)
4-Cyano-4'-hexylbiphenyl
Storage condition: Sealed in dry, Room Temperature
Formula:
C₁₉H₂₁N Storage Temperature: Ambient |
MDL Number:
MFCD00075144 CAS Number: 41122-70-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...