4-(Methylthio)benzylamine 98+%

BD91439-1G BD91439-100MG
BLDPBD91439-1GEA 58.8 EUR
BLDPBD91439-1G BLDPBD91439-100MG
4-(Methylthio)benzylamine 98+%
4-(Methylthio)benzylamine
Formula: C₈H₁₁NS
MDL Number: MFCD01310831
CAS Number: 83171-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR