4-(4-Morpholinylcarbonyl)phenylboronic acid pinacol ester 98%
|
|
BD91452-100MG
BD91452-5G
BLDPBD91452-100MGEA
18.9
EUR
BLDPBD91452-100MG
BLDPBD91452-5G
4-(4-Morpholinylcarbonyl)phenylboronic acid pinacol ester 98%
4-(4-Morpholinylcarbonyl)phenylboronic acid pinacol ester
Storage condition: Sealed in dry, Room Temperature
Formula:
C₁₇H₂₄BNO₄ MW: 317.19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD05863913 CAS Number: 656239-38-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...